Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Heptylphenol 98.0+%, TCI America™
SDP

Catalog No. H061525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H061525G 25 g
H06155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H061525G Supplier TCI America Supplier No. H061525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1987-50-4
Molecular Formula C13H20O
Molecular Weight (g/mol) 192.302
MDL Number MFCD00041751
InChI Key KNDDEFBFJLKPFE-UHFFFAOYSA-N
Synonym 4-n-heptylphenol, phenol, 4-heptyl, p-heptylphenol, heptanyl-p-phenol, p-hydroxyheptylbenzene, phenol, heptyl derivs., p-n-heptylphenol, unii-i743z35dvb, phenol, heptenylated, p-heptyl phenol
PubChem CID 16143
ChEBI CHEBI:34438
IUPAC Name 4-heptylphenol
SMILES CCCCCCCC1=CC=C(C=C1)O

Specifications

Color Red-White
Quantity 25 g
UN Number 2430
Formula Weight 192.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Heptylphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.