Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. H16025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H16025G 5 g
H16021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H16025G Supplier TCI America Supplier No. H16025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2836-44-4
Molecular Formula C6H5F3N2O
Molecular Weight (g/mol) 178.114
MDL Number MFCD03265321
InChI Key DBDRDWKBAGNHHS-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl pyrimidin-4-ol, 4-hydroxy-2-methyl-6-trifluoromethylpyrimidine, 4-hydroxy-2-methyl-6-trifluoromethyl pyrimidine, 4-pyrimidinol, 2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethylpyrimidin-4-ol, 2-methyl-6-trifluoromethyl-4-pyrimidinol, 2-methyl-4-hydroxy-6-trifluoromethylpyrimidine, 2-methyl-6-trifluoromethyl-1h-pyrimidin-4-one, 2-methyl-6-trifluoromethyl pyrimidin-4 3h-one, 4 1h-pyrimidinone, 2-methyl-6-trifluoromethyl
PubChem CID 233901
IUPAC Name 2-methyl-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES CC1=NC(=O)C=C(N1)C(F)(F)F

Specifications

Melting Point 143°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-2-methyl-6-(trifluoromethyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.