Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxy-alpha-methylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. H13861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13861G 1 g
H13865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13861G Supplier TCI America Supplier No. H13861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2480-86-6
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00238638
InChI Key BDRRAMWDUCXAKG-UHFFFAOYNA-N
Synonym 2-Hydroxy-5-(1-hydroxyethyl)anisole, 4-(1-Hydroxyethyl)-2-methoxyphenol
PubChem CID 17203
ChEBI CHEBI:86590
IUPAC Name 4-(1-hydroxyethyl)-2-methoxyphenol
SMILES COC1=CC(=CC=C1O)C(C)O

Specifications

Melting Point 102°C
Quantity 1 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxy-alpha-methylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.