Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-methoxycinnamic acid, 99%

Catalog No. AC156360250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156360250 25 g Glass bottle
AC156360050 5 g Glass bottle
AC156361000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156360250 Supplier Thermo Scientific Chemicals Supplier No. 156360250
  • $120.00
    $95.65 / Each of 1

    Save $24.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1135-24-6 | C10H10O4 | 194.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-24-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00004400
InChI Key KSEBMYQBYZTDHS-HWKANZROSA-N
Synonym ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
PubChem CID 445858
ChEBI CHEBI:17620
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=CC(=C1)C=CC(=O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 169.0°C to 173.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)CH=CHCO2H
Quantity 25 g
Beilstein 10, 436
Merck Index 15, 4089
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ethyl acetate,moderately soluble in diethyl ether,sparingly soluble in petroleum ether and benzene
Formula Weight 194.19
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-3-methoxycinnamic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-490-0
RTECSNumber UD3365500
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.