Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-nitroso-1-naphthalenesulfonic Acid Hydrate 98.0+%, TCI America™
SDP

Catalog No. N026925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026925G 25 g
N02695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N026925G Supplier TCI America Supplier No. N026925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3682-32-4
Molecular Formula C10H7NO5S
Molecular Weight (g/mol) 253.228
MDL Number MFCD00003933
InChI Key GASPSNIEUBWWJU-UHFFFAOYSA-N
Synonym 2-nitroso-1-naphthol-4-sulfonic acid, 4-hydroxy-3-nitroso-1-naphthalenesulfonic acid, 1-naphthalenesulfonic acid, 4-hydroxy-3-nitroso, unii-7t06pca43o, 4-hydroxy-3-nitroso-naphthalene-1-sulfonic acid, 4-hydroxy-3-nitrosonaphthalenesulfonic acid, nitroso-nw acid, acmc-1cril, 4-hydroxy-3-nitrosonaphthalene-1-sulphonic acid, 1-naphthalenesulfonicacid, 4-hydroxy-3-nitroso
PubChem CID 77257
IUPAC Name 4-hydroxy-3-nitrosonaphthalene-1-sulfonic acid
SMILES C1=CC=C2C(=C1)C(=CC(=C2O)N=O)S(=O)(=O)O

Specifications

Melting Point 145°C
Quantity 25 g
Formula Weight 253.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-nitroso-1-naphthalenesulfonic Acid Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.