Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-4'-nitrostilbene, 97%, Thermo Scientific™

Catalog No. AC450890050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC450890050 Supplier Thermo Scientific Chemicals Supplier No. 450890050

Chemical Identifiers

CAS 19221-08-0
Molecular Formula C14H11NO3
Molecular Weight (g/mol) 241.25
InChI Key OETQWIHJPIESQB-OWOJBTEDSA-N
Synonym 4-hydroxy-4'-nitrostilbene, 4-4-nitrostyryl phenol, 4-e-2-4-nitrophenyl ethenyl phenol, 4-hydroxy-4/'-nitrostilbene, 4-1e-2-4-nitrophenyl vinyl phenol, 4'-nitrostilben-4-ol, cambridge id 5309411, e-4'-nitrostilbene-4-ol, e-4-4-nitrostyryl phenol
PubChem CID 759250
IUPAC Name 4-[(E)-2-(4-nitrophenyl)ethenyl]phenol
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)O)[N+](=O)[O-]

Specifications

Melting Point 209.0°C to 210.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 241.25
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-4'-nitrostilbene
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.