Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-6-methyl-2-pyrone 98.0+%, TCI America™
SDP

Catalog No. H0715500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0715500G 500 g
H071525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0715500G Supplier TCI America Supplier No. H0715500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 675-10-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
MDL Number MFCD00006641,MFCD18820248
InChI Key OOKCZXGEYPSNIM-UHFFFAOYSA-N
Synonym 4-hydroxy-6-methyl-2-pyrone, 2-hydroxy-6-methyl-4h-pyran-4-one, 4-hydroxy-6-methyl-2h-pyran-2-one, triacetic acid lactone, triacetate lactone, 2h-pyran-2-one, 4-hydroxy-6-methyl, 6-methyl-4-hydroxypyron-2, unii-s1s883s4ee, 6-methyl-4-hydroxy-2-pyrone, ccris 3600
PubChem CID 54675757
ChEBI CHEBI:16458
IUPAC Name 2-hydroxy-6-methyl-4H-pyran-4-one
SMILES CC1=CC(=O)C=C(O)O1

Specifications

Melting Point 188°C
Quantity 500 g
Formula Weight 126.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-6-methyl-2-pyrone
RTECSNumber UQ1000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.