Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-L-(+)-2-phenylglycine 99.0+%, TCI America™
SDP

Catalog No. H138925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H138925G 25 g
H13895G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H138925G Supplier TCI America Supplier No. H138925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32462-30-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00065932
InChI Key LJCWONGJFPCTTL-ZETCQYMHSA-N
Synonym (S)-(+)-2-Amino-2-(4-hydroxyphenyl)acetic Acid, L-(+)-2-(4-Hydroxyphenyl)glycine, Oxfenicine, H-Phg(4-OH)-OH
PubChem CID 36143
ChEBI CHEBI:31755
IUPAC Name (2S)-2-amino-2-(4-hydroxyphenyl)acetic acid
SMILES C1=CC(=CC=C1C(C(=O)O)N)O

Specifications

Melting Point 214°C
Color White-Yellow
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxy-L-(+)-2-phenylglycine
EINECSNumber (3)-3667
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.