Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzamide 98.0+%, TCI America™
SDP

Catalog No. H020025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H020025G 25 g
H02005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H020025G Supplier TCI America Supplier No. H020025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 619-57-8
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00007997
InChI Key QXSAKPUBHTZHKW-UHFFFAOYSA-N
Synonym benzamide, 4-hydroxy, 4-hydroxy-benzamide, p-hydroxybenzamide, unii-9ou5yd093j, 4-hydroxy benzamide, 1,3,4,9-tetrahydro-2-hydroxybenzoyl-9-4-hydroxyphenyl methyl-6-methoxy-2h-pyrido 3,4-b indole, 4-oxidanylbenzamide, acmc-1apvc, para-hydroxybenzoic amide
PubChem CID 65052
IUPAC Name 4-hydroxybenzamide
SMILES NC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 161°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.