Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzenesulfonamide 97.0+%, TCI America™
SDP

Catalog No. H0328250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0328250G 250 g
H032825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0328250G Supplier TCI America Supplier No. H0328250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1576-43-8
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 173.186
MDL Number MFCD00059190
InChI Key DIRCLGLKRZLKHG-UHFFFAOYSA-N
Synonym p-hydroxybenzenesulfonamide, p-phenolsulfonamide, benzenesulfonamide, 4-hydroxy, 4-sulfamoylphenol, benzenesulfonamide, p-hydroxy, 4-hydroxyphenylsulfonamide, unii-1iit7r7vpj, p-hydroxybenzenesulphonamide, 4-hydroxybenzenesulphonamide, 4-hydroxybenzene-1-sulfonamide
PubChem CID 74093
IUPAC Name 4-hydroxybenzenesulfonamide
SMILES C1=CC(=CC=C1O)S(=O)(=O)N

Specifications

Melting Point 174°C
Quantity 250 g
Formula Weight 173.19
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.