Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzoic acid, 99%

Catalog No. AAA137000B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA137000B 1000 g
AAA1370030 250 g
AAA137000I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA137000B Supplier Thermo Scientific Chemicals Supplier No. A137000B
Add to Cart

CAS: 99-96-7 | C7H6O3 | 138.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Hydroxybenzoic acid is used as an intermediate for dyes, antiseptics and active pharmaceutical ingredients. It is also used as a food preservative, corrosion inhibitor, anti-oxidant and an emulsifier. It reacts with 6-hydroxynaphthalene-2-carboxylic acid to prepare aromatic polyester called Vectran fiber. It acts as a precursor to prepare p-hydroxyphenylbenzoate, p-acetoxybenzoyl chloride and 4,4′-dihydroxybenzophenone. Further, it is used in paint additives, coating additives, processing aids, electrical and electronic products. In addition to this, it acts as a preservative in cosmetics and some ophthalmic solutions.

Solubility
Soluble in water, alcohol, ethanol, ether, n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-96-7
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00002547
InChI Key FJKROLUGYXJWQN-UHFFFAOYSA-N
Synonym p-hydroxybenzoic acid, 4-carboxyphenol, benzoic acid, 4-hydroxy, p-salicylic acid, benzoic acid, p-hydroxy, para-hydroxybenzoic acid, 4-hydroxybenzoesaeure, p-carboxyphenol, 4-hydroxy benzoic acid, p-oxybenzoesaure german
PubChem CID 135
ChEBI CHEBI:30763
IUPAC Name 4-hydroxybenzoic acid
SMILES OC(=O)C1=CC=C(O)C=C1

Specifications

Melting Point 213°C to 218°C
Flash Point 199°C (390°F)
Assay Percent Range 99%
Quantity 1000 g
Beilstein 970950
Merck Index 14,4812
Solubility Information Soluble in water,alcohol,ethanol,ether,n-butanol and acetone. Slightly soluble in chloroform. Insoluble in carbon disulfide.
Formula Weight 138.12
Percent Purity 99%
Chemical Name or Material 4-Hydroxybenzoic acid
Hazard Category H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c
EINECSNumber 202-804-9
RTECSNumber DH1925000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.