Learn More
4-Hydroxybenzonitrile, 98+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
4-Hydroxybenzonitrile is used as an intermediate in organic synthesis. It is also used in cosmetic as an antiseptic and disinfectant. It is involved as a raw material for the preparation of bomoxynil and ioxynil.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 110°C to 114°C |
| Quantity | 100 g |
| Beilstein | 386130 |
| Solubility Information | Soluble in methanol. Insoluble in water. |
| Formula Weight | 119.12 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 4-Hydroxybenzonitrile |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| EINECSNumber | 212-175-2 |
| RTECSNumber | DI4375000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.