Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. H0224100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0224100G 100 g
H022425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0224100G Supplier TCI America Supplier No. H0224100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00004658
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O

Specifications

Color White-Yellow
Quantity 100 g
Formula Weight 124.14
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzyl Alcohol
EINECSNumber (3)-1052
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.