Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzyl Cyanide 99.0+%, TCI America™
SDP

Catalog No. H0657500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0657500G 500 g
H065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0657500G Supplier TCI America Supplier No. H0657500G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 14191-95-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00002383
InChI Key AYKYOOPFBCOXSL-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl cyanide, 4-hydroxyphenylacetonitrile, 2-4-hydroxyphenyl acetonitrile, p-hydroxybenzyl cyanide, 4-hydroxyphenyl acetonitrile, p-hydroxyphenylacetonitrile, 4-hydroxybenzylcyanide, 4-hydroxybenzeneacetonitrile, benzeneacetonitrile, 4-hydroxy, acetonitrile, p-hydroxyphenyl
PubChem CID 26548
ChEBI CHEBI:16667
IUPAC Name 2-(4-hydroxyphenyl)acetonitrile
SMILES OC1=CC=C(CC#N)C=C1

Specifications

Melting Point 71°C
Boiling Point 150°C
Quantity 500 g
Formula Weight 133.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxybenzyl Cyanide
RTECSNumber AM0530000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.