Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxybenzylamine hydrate, 97%

Catalog No. AC432930250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432930250 25 g Glass bottle
AC432930050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432930250 Supplier Thermo Scientific Chemicals Supplier No. 432930250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 696-60-6 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-60-6
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD19690971
InChI Key RQJDUEKERVZLLU-UHFFFAOYSA-N
Synonym 4-hydroxybenzylamine, 4-aminomethyl phenol, 4-aminomethyl-phenol, para-hydroxybenzylamine, unii-7j7f85b7bi, 4-hydroxyphenyl methanaminium bromide, a-amino-p-cresol, 4-hyroxybenzylamine, p-hydroxybenzylamine, 4-hydroxylbenzylamine
PubChem CID 97472
IUPAC Name 4-(aminomethyl)phenol
SMILES NCC1=CC=C(O)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 100°C to 104°C
Color White to Yellow
Elemental Analysis 57.18 to 61.94% C, 9.52 to 10.31% N (based on monohydrate)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 141.17
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Hydroxybenzylamine hydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.