Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxycarbazole 98.0+%, TCI America™
SDP

Catalog No. H101425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H101425G 25 g
H10141G 1 g
H10145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H101425G Supplier TCI America Supplier No. H101425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 52602-39-8
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD02178385
InChI Key UEOHATPGKDSULR-UHFFFAOYSA-N
Synonym 4-hydroxycarbazole, 4-hydroxy carbazole, ccris 5300, chembl46723, 4-hydroxycarbazol, 4-hydroxy-carbazole, pubchem9190, 4-hydroxy-9h-carbazole, acmc-209l0g
PubChem CID 104251
IUPAC Name 9H-carbazol-4-ol
SMILES C1=CC=C2C(=C1)C3=C(N2)C=CC=C3O

Specifications

Melting Point 169°C
Quantity 25 g
Formula Weight 183.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxycarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.