Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxycoumarin 98.0+%, TCI America™
SDP

Catalog No. H0235250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0235250G 250 g
H023525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0235250G Supplier TCI America Supplier No. H0235250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key OWBBAPRUYLEWRR-UHFFFAOYSA-N
Synonym 4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
IUPAC Name 2-hydroxy-4H-chromen-4-one
SMILES OC1=CC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 212°C
Color Yellow
Quantity 250 g
Formula Weight 162.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxycoumarin
RTECSNumber DJ3100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.