Learn More
Abcam 4-Hydroxydiclofenac, Diclofenac Metabolite, Purity >97%, 10 mg
Description
- Active 4'-hydroxylated metabolite of diclofenac
- Suppresses prostaglandin E2 (PGE2) formation by specifically blocking COX2 activity
- Demonstrates anti-inflammatory and analgesic properties
- Molecular weight: 312.1 g/mol
- Purity: >97%
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Product Type | Antioxidant |
| Target Molecule | COX2 |
| Quantity | 10 mg |
| For Use With (Application) | COX2-specific inhibition |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.