Learn More
Medchemexpress LLC 4-hydroxyindole-3-acetic acid | 56395-08-5 | MFCD01326488 | 99.1% | 191.18 | C10H9NO3 | 25 MG

Supplier: Medchemexpress LLC HYW50938125MG
4-Hydroxyindole-3-acetic acid (4HIAA) is a research-grade drug metabolite and analytical standard supplied as an off-white to light yellow solid. It is the primary oxidative metabolite of psilocybin and psilocin and is intended for analytical and metabolic studies. Store under nitrogen at recommended temperatures to preserve stability.
- High purity (99.13% by HPLC).
- Molecular formula C10H9NO3; molecular weight 191.18.
- Available in small research sizes: 5 MG, 10 MG, 25 MG.
- Appearance: off-white to light yellow solid.
- Storage: 4°C under nitrogen; in solvent keep at -80°C up to 6 months or -20°C up to 1 month.
- For research/analytical use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.