Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyindole, 98%

Catalog No. AAL14352ME
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL14352ME 500 mg
AAL1435203 1 g
AAL1435206 5 g
AAL1435214 25 g
AAL14352MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAL14352ME Supplier Thermo Scientific Chemicals Supplier No. L14352ME

CAS: 2380-94-1 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-94-1
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005667
InChI Key NLMQHXUGJIAKTH-UHFFFAOYSA-N
Synonym 4-hydroxyindole, indol-4-ol, 4-indolol, hydroxyindole, 4-hydroxy-1h-indole, 1h-indole-4-ol, 4-hydroxy indole, unii-1w4vd9085v, hydroxyindoles, 4-hydroxy-indole
PubChem CID 75421
ChEBI CHEBI:24702
IUPAC Name 1H-indol-4-ol
SMILES OC1=C2C=CNC2=CC=C1

Specifications

Melting Point 97°C to 99°C
Quantity 500 mg
Beilstein 114905
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxyindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-177-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.