Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxynonenal Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3249
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3249 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3249 Supplier R&D Systems Supplier No. MAB3249
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 24 publications

4-Hydroxynonenal Monoclonal specifically detects 4-Hydroxynonenal in Multi-Species samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen 4-Hydroxynonenal
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 198960
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 0.1-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Alias (2E)-4-Hydroxy-2-nonenal, 2-Nonenal, 4-hydroxy-, (2E,4R)-, 4 HNE, 4Hydroxynonenal, 4-hydroxy-trans-2-nonenal, HNE
Host Species Mouse
Immunogen KLH-coupled 4-Hydroxynonenal
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects 4-Hydroxynonenal adducts of histidine residues. No cross-reactivity with nitrotyrosine, formaldehyde, glutaraldehyde, or 4-hydroxy hexenaldehyde adducts is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.