Learn More
Abcam 4-Hydroxynonenal, Lipid Peroxidation Marker, Liquid, 1 mg
Description
- Purity: >97%
- Molecular Weight: 156.22 Da
- Form: Liquid
- Marker of lipid peroxidation: Prevents NF-κB activation and TNF expression by inhibiting IκB phosphorylation
- Biological activity: Cytotoxicity, growth inhibiting activity, genotoxicity, and chemotactic activity; active in vivo
- Storage buffer: Constituents: 100% (E)-4-hydroxynon-2-enal
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | 4-Hydroxynonenal |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Buffer | 100% (E)-4-hydroxynon-2-enal |
| Form | Liquid |
| Content And Storage | Store at -20°C. |
| Formulation | 100% (E)-4-hydroxynon-2-enal |
| Quantity | 1 mg |
| For Use With (Application) | Lipid peroxidation marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.