Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-Hydroxynonenal Monoclonal Antibody (12F7), PerCP
GREENER_CHOICE

Catalog No. PIMA545791
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545791 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545791 Supplier Invitrogen™ Supplier No. MA545791
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

A 1:1,000 dilution of MA5-45791 was sufficient for detection of 4-Hydroxynonenal in 0.5 μg of 4-Hydroxynonenal conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody. Specific for 4-Hydroxynonenal (4-HNE) modified proteins. Does not detect free 4-Hydroxynonenal. Does not cross-react with 4-Hydroxy-2-hexenal, Acrolein, Crotonaldehyde, Hexanoyl Lysine, Malondialdehyde, or Methylglyoxal modified proteins.

4-Hydroxynonenal, or 4-hydroxy-2-nonenal or 4-HNE or HNE, is an alpha, beta-unsaturated hydroxyalkenal that is produced by lipid peroxidation in cells. 4-HNE is the primary alpha, beta-unsaturated hydroxyalkenal formed in this process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Hydroxynonenal
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 12F7
Concentration 1 mg/mL
Conjugate PerCP
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.4
Gene Alias 4HNE; 4-HNE; 4-hydroxy Hexenal; 4-Hydroxy Nonenal; 4-hydroxy-2-nonenal; 4-Hydroxynonenal; HNE
Host Species Mouse
Immunogen Synthetic 4-Hydroxynonenal modified Keyhole Limpet Hemocyanin (KLH).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.