Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Hydroxyphenoxy)acetic Acid 99.0+%, TCI America™
SDP

Catalog No. H099225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H099225G 25 g
H09925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H099225G Supplier TCI America Supplier No. H099225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1878-84-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00014362
InChI Key PKGWLCZTTHWKIZ-UHFFFAOYSA-N
Synonym 4-hydroxyphenoxyacetic acid, 4-hydroxyphenoxy acetic acid, 2-4-hydroxyphenoxy acetic acid, 4-hydroxypheoxyacetate, acetic acid, 4-hydroxyphenoxy, 4-hydroxy-phenoxy-acetic acid, p-hydroxyphenoxyacetic acid, acetic acid, 2-4-hydroxyphenoxy, acetic acid, p-hydroxyphenoxy, 4-carboxymethoxy phenol
PubChem CID 15881
ChEBI CHEBI:1881
IUPAC Name 2-(4-hydroxyphenoxy)acetic acid
SMILES C1=CC(=CC=C1O)OCC(=O)O

Specifications

Melting Point 155°C
Color White-Yellow
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (4-Hydroxyphenoxy)acetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.