Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypyridine-2-carboxylic acid, 97%

Catalog No. AAH3426203
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3426203 1 g
AAH34262MD 250 mg
AAH3426206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3426203 Supplier Thermo Scientific Chemicals Supplier No. H3426203
Add to Cart

CAS: 22468-26-4 | C6H5NO3 | 139.11 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
slightly soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22468-26-4
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00521155,MFCD11656334
InChI Key MXXLHBCSVDDTIX-UHFFFAOYSA-N
Synonym 4-hydroxypicolinic acid, 4-hydroxypyridine-2-carboxylic acid, 4-hydroxy-2-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxypyridine-2-carboxylicacid, 2-pyridinecarboxylic acid, 1,4-dihydro-4-oxo, 4-hydroxy-picolinsaure, pubchem17131, acmc-209fwn
PubChem CID 735149
IUPAC Name 4-oxo-1H-pyridine-2-carboxylic acid
SMILES OC(=O)C1=CC(=O)C=CN1

Specifications

Melting Point ∽280°C (decomposition)
Quantity 1 g
Solubility Information slightly soluble in water.
Formula Weight 139.11
Percent Purity 97%
Chemical Name or Material 4-Hydroxypyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.