Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxypyridine, 97%

Catalog No. AAA1458622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1458622 100 g
AAA1458614 25 g
AAA1458636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1458622 Supplier Thermo Scientific Chemicals Supplier No. A1458622
Add to Cart

CAS: 626-64-2 | C5H5NO | 95.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-64-2
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006419,MFCD00040458
InChI Key GCNTZFIIOFTKIY-UHFFFAOYSA-N
Synonym 4-hydroxypyridine, pyridin-4-ol, 4-pyridinol, 4-pyridone, 4 1h-pyridinone, pyridin-4 1h-one, 4-oxopyridine, 4-pyridinone, 4-pyridol, gamma-hydroxypyridine
PubChem CID 12290
IUPAC Name 1H-pyridin-4-one
SMILES O=C1C=CNC=C1

Specifications

Melting Point 142°C to 148°C
Boiling Point 230°C to 235°C (12 mmHg)
Quantity 100 g
Beilstein 105800
Sensitivity Hygroscopic; Light sensitive
Formula Weight 95.1
Percent Purity 97%
Chemical Name or Material 4-Hydroxypyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-958-3
RTECSNumber UU7701450
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.