Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxyquinoline, 98%

Catalog No. AAA1585906
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1585906 5 g
AAA1585903 1 g
AAA1585909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1585906 Supplier Thermo Scientific Chemicals Supplier No. A1585906
Add to Cart

CAS: 611-36-9 | C9H7NO | 145.16 g/mol

4-Hydroxyquinoline is a medical raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxyquinoline is a medical raw material.

Solubility
Soluble in methanol.

Notes
Store at room temperature. Keep away from oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-36-9
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006777,MFCD00956391
InChI Key PMZDQRJGMBOQBF-UHFFFAOYSA-N
Synonym 4-hydroxyquinoline, quinolin-4-ol, 4-quinolinol, quinolin-4 1h-one, 4 1h-quinolinone, kynurine, 4-hydroxy quinoline, quinoline, 4-hydroxy, ccris 4329, 4-chinolinol
PubChem CID 69141
ChEBI CHEBI:15815
SMILES O=C1C=CNC2=CC=CC=C12

Specifications

Melting Point 201°C to 205°C
Quantity 5 g
Beilstein 1524969
Solubility Information Soluble in methanol.
IUPAC Name 1,4-dihydroquinolin-4-one
Formula Weight 145.16
Percent Purity 98%
Chemical Name or Material 4-Hydroxyquinoline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-268-2
RTECSNumber VC4070000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.