Learn More
BIOGEMS INTERNATIONAL INC 4-Hydroxytamoxifen 50mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 683358550MG
Encompass_Preferred
Tamoxifen is a modulator of the of the estrogen receptor, with tissue specific agonistic and antagonistic effects. 4-hydroxy Tamoxifen is the active metabolite of tamoxifen that is also generated in the liver by the CYP2D6 enzyme. It exhibits a greater potency than the parent compound and is reported to be cytostatic and prevent cancerous cells from dividing.
Catalog No. 50-153-0236
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.