Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-2-methoxypyridine-3-carboxaldehyde, 95%, Thermo Scientific Chemicals

Catalog No. AAH6698403
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6698403 1 g
AAH66984MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6698403 Supplier Thermo Scientific Chemicals Supplier No. H6698403
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 158669-26-2
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD05662396
InChI Key GNBKAOHTTIVAMT-UHFFFAOYSA-N
Synonym 4-iodo-2-methoxynicotinaldehyde, 4-iodo-2-methoxypyridine-3-carboxaldehyde, 3-formyl-4-iodo-2-methoxypyridine, 4-iodo-2-methoxy-pyridine-3-carbaldehyde, 3-pyridinecarboxaldehyde, 4-iodo-2-methoxy, 2-methoxy-4-iodo-3-pyridinecarboxyaldehyde, pubchem19522, iodomethoxynicotinaldehyde, 2-methoxy-4-iodopyridine-3-carbaldehyde, 4-iodo-2-methoxy-3-pyridinecarboxaldehyde
PubChem CID 10038381
IUPAC Name 4-iodo-2-methoxypyridine-3-carbaldehyde
SMILES COC1=NC=CC(I)=C1C=O

Specifications

Quantity 1 g
Beilstein 7020882
Sensitivity Light sensitive
Formula Weight 263.03
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Iodo-2-methoxypyridine-3-carboxaldehyde
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.