Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-4',4″-dimethoxytriphenylamine 98.0+%, TCI America™
SDP

Catalog No. I07765G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07765G 5 g
I07761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07765G Supplier TCI America Supplier No. I07765G

Chemical Identifiers

CAS 201802-15-5
Molecular Formula C20H18INO2
Molecular Weight (g/mol) 431.273
InChI Key VGQDQJMYUYSINZ-UHFFFAOYSA-N
Synonym 4-Iodo-N,N-bis(4-methoxyphenyl)aniline
PubChem CID 44630018
IUPAC Name N-(4-iodophenyl)-4-methoxy-N-(4-methoxyphenyl)aniline
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)OC)C3=CC=C(C=C3)I

Specifications

Melting Point 111°C
Quantity 5 g
Formula Weight 431.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-4′,4′′-dimethoxytriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.