Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-4'-propylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. I10101G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10101G 1 g
I1010200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10101G Supplier TCI America Supplier No. I10101G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 782477-81-0
Molecular Formula C15H15I
Molecular Weight (g/mol) 322.19
MDL Number MFCD12923241
InChI Key MKDLYSABBHXXSV-UHFFFAOYSA-N
PubChem CID 53401151
IUPAC Name 4-iodo-4'-propyl-1,1'-biphenyl
SMILES CCCC1=CC=C(C=C1)C1=CC=C(I)C=C1

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 1 g
Formula Weight 322.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Iodo-4′-propylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.