Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodo-o-xylene, 98+%

Catalog No. AAA1323218
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1323218 50 g
AAA1323209 10 g
AAA1323230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1323218 Supplier Thermo Scientific Chemicals Supplier No. A1323218
Add to Cart

CAS: 31599-61-8 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31599-61-8
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00040989
InChI Key CSFRCLYFVINMBZ-UHFFFAOYSA-N
Synonym 4-iodo-o-xylene, 1,2-dimethyl-4-iodobenzene, benzene, 4-iodo-1,2-dimethyl, 3,4-dimethyliodobenzene, 1-iodo-3,4-dimethylbenzene, 4-iodo-ortho-xylene, 3,4-dimethyl-1-iodobenzene, 4-iodo-orthoxylene, 4-iodo-0-xylene, pubchem3105
PubChem CID 141646
IUPAC Name 4-iodo-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)I)C

Specifications

Density 1.633
Quantity 50 g
Refractive Index 1.602
Beilstein 1905729
Boiling Point 231°C to 233°C
Formula Weight 232.07
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-o-xylene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.