Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodoaniline, 99%

Catalog No. AC156615000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156615000 500 g Glass bottle
AC156610250 25 g Glass bottle
AC156611000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC156615000 Supplier Thermo Scientific Chemicals Supplier No. 156615000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 540-37-4 | C6H6IN | 219.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-37-4
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.02
InChI Key VLVCDUSVTXIWGW-UHFFFAOYSA-N
Synonym p-iodoaniline, benzenamine, 4-iodo, 4-iodobenzenamine, aniline, p-iodo, p-aminophenyl iodide, aniline, 4-iodo, 4-iodo-aniline, 4-iodophenylamine, 4-iodo-benzenamine, 4-iodo-phenylamine
PubChem CID 10893
IUPAC Name 4-iodoaniline
SMILES C1=CC(=CC=C1N)I

Specifications

Melting Point 59.0°C to 64.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4NH2
Quantity 500 g
Beilstein 12,67
Merck Index 15,507
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,chloroform and ether
Formula Weight 219.02
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 4-Iodoaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 208-743-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.