Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Iodophenetole 98.0+%, TCI America™
SDP

Catalog No. I052525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I052525G 25 g
I05255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I052525G Supplier TCI America Supplier No. I052525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 699-08-1
Molecular Formula C8H9IO
Molecular Weight (g/mol) 248.063
MDL Number MFCD00061124
InChI Key VSIIHWOJPSSIDI-UHFFFAOYSA-N
Synonym 4-iodophenetole, p-iodophenetole, 1-ethoxy-4-iodo-benzene, 4-ethoxyiodobenzene, 4-ethoxy-4-iodobenzene, benzene, 1-ethoxy-4-iodo, 4-iodophenyl ethyl ether, phenetole, p-iodo, p-ethoxyiodobenzene, ethyl 4-iodophenyl ether
PubChem CID 220700
IUPAC Name 1-ethoxy-4-iodobenzene
SMILES CCOC1=CC=C(C=C1)I

Specifications

Quantity 25 g
Formula Weight 248.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Iodophenetole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.