Learn More
Selleck Chemical LLC 4-IPP S2643-5mg
SDP
Supplier:  Selleck Chemical LLC S26435MG
Encompass_Preferred
4-IPP (4-Iodo-6-phenylpyrimidine) is an irreversible inhibitor of macrophage migration inhibitory factor (MIF) and a specific suicide substrate for MIF 4-IPP can inhibit receptor activator of NF- B ligand (RANKL)-induced osteoclastogenesis and potentiate osteoblast-mediated mineralization and bone nodule formation in vitro
Catalog No. 50-217-3093
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.