Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isobutoxy-2-butanone 95.0+%, TCI America™
SDP

Catalog No. I02865ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I02865ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I02865ML Supplier TCI America Supplier No. I02865ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31576-33-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00059348
InChI Key VDVQAHFUWSAGRT-UHFFFAOYSA-N
PubChem CID 537792
IUPAC Name 4-(2-methylpropoxy)butan-2-one
SMILES CC(C)COCCC(=O)C

Specifications

Color Yellow
Boiling Point 181°C
Quantity 5 mL
Formula Weight 144.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isobutoxy-2-butanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.