Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isobutylbenzaldehyde 95.0+%, TCI America™
SDP

Catalog No. I0436100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I0436100ML 100 mL
I043625ML 25 mL
I0436500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. I0436100ML Supplier TCI America Supplier No. I0436100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40150-98-9
Molecular Formula C11H14O
Molecular Weight (g/mol) 162.232
MDL Number MFCD00075760
InChI Key LXPWGAZYJHUWPM-UHFFFAOYSA-N
PubChem CID 577537
IUPAC Name 4-(2-methylpropyl)benzaldehyde
SMILES CC(C)CC1=CC=C(C=C1)C=O

Specifications

Color Yellow
Quantity 100 mL
Formula Weight 162.23
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isobutylbenzaldehyde
EINECSNumber (3)-2663
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.