Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropenyltoluene (stabilized with TBC) 95.0+%, TCI America™
SDP

Catalog No. I069325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I069325G 25 g
I06935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I069325G Supplier TCI America Supplier No. I069325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1195-32-0
Molecular Formula C10H12
Molecular Weight (g/mol) 132.21
MDL Number MFCD00036510
InChI Key MMSLOZQEMPDGPI-UHFFFAOYSA-N
Synonym 1-methyl-4-prop-1-en-2-yl benzene, dehydro-p-cymene, 2-p-tolylpropene, 4-isopropenyltoluene, 4-methylisopropenylbenzene, 2-p-methylphenyl propene, p-cymenene, p,alpha-dimethylstyrene, p-alpha-dimethylstyrene, 1-isopropenyl-4-methylbenzene
PubChem CID 62385
IUPAC Name 1-methyl-4-(prop-1-en-2-yl)benzene
SMILES CC(=C)C1=CC=C(C)C=C1

Specifications

Melting Point -28°C
Color Yellow
Boiling Point 186°C
Quantity 25 g
Formula Weight 132.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Isopropenyltoluene (stabilized with TBC)
EINECSNumber (3)-0008&(3)-0010
TSCA Yes
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.