Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropoxybenzenesulfonyl chloride, 96%

Catalog No. AAL1898414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1898414 25 g
AAL1898403 1 g
AAL1898406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1898414 Supplier Thermo Scientific Chemicals Supplier No. L1898414

CAS: 98995-40-5 | C9H11ClO3S | 234.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98995-40-5
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD03093083
InChI Key IJWCRHKAQNFJLT-UHFFFAOYSA-N
Synonym 4-isopropoxybenzenesulfonyl chloride, 4-isopropoxybenzene-1-sulfonyl chloride, 4-isopropoxyphenylsulfonyl chloride, 4-propan-2-yloxy benzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy, 4-isopropoxybenzenesulphonyl chloride, benzenesulfonyl chloride, 4-1-methylethoxy-9ci, 4-isopropoxybenzenesulfonylchloride, pubchem19078, acmc-209sbk
PubChem CID 22327297
IUPAC Name 4-propan-2-yloxybenzenesulfonyl chloride
SMILES CC(C)OC1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 30°C to 37°C
Boiling Point 112°C to 113°C (0.01 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 3269058
Sensitivity Moisture sensitive
Formula Weight 234.7
Percent Purity 96%
Chemical Name or Material 4-Isopropoxybenzenesulfonyl chloride, 25g
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.