Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylbenzonitrile, 95%, Thermo Scientific™

Catalog No. SB00932DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB00932DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB00932DE Supplier Thermo Scientific Chemicals Supplier No. SB00932DE
Add to Cart

Chemical Identifiers

CAS 13816-33-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00052966
InChI Key YFDJCWXBKWRDPW-UHFFFAOYSA-N
Synonym 4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
PubChem CID 26289
IUPAC Name 4-propan-2-ylbenzonitrile
SMILES CC(C)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula (CH3)2CHC6H4CN
Quantity 5 g
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 4-Isopropylbenzonitrile
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.