Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 4-Isopropylbenzyl alcohol, 97+%

Catalog No. AAH3669222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3669222 100 g
AAH3669214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3669222 Supplier Thermo Scientific™ Supplier No. H3669222
  • $93.60
    $79.65 / Each of 1

    Save $13.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 536-60-7 | C10H14O | 150.22 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 536-60-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00004663
InChI Key OIGWAXDAPKFNCQ-UHFFFAOYSA-N
Synonym Cumic alcohol;
IUPAC Name [4-(propan-2-yl)phenyl]methanol
SMILES CC(C)C1=CC=C(CO)C=C1

Specifications

Density 0.980
Boiling Point 135°C to 136°C/26 mm
Flash Point >110°C(230°F)
Quantity 100 g
Merck Index 14,2619
Formula Weight 150.22
Percent Purity 97+%
Physical Form Liquid
ShelfLife 5 Years
EINECSNumber 208-640-4
RTECSNumber GZ7260000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.