Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Isopropylcarbamoyl)benzeneboronic acid, 95%

Catalog No. AAH5292503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5292503 1 g
AAH52925MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5292503 Supplier Thermo Scientific Chemicals Supplier No. H5292503
Add to Cart

CAS: 397843-67-3 | C10H14BNO3 | 207.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397843-67-3
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.036
MDL Number MFCD04039353
InChI Key GBCSEYKTZAKRMT-UHFFFAOYSA-N
Synonym 4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid
PubChem CID 2773475
IUPAC Name [4-(propan-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O

Specifications

Melting Point 162°C to 167°C
Quantity 1 g
Formula Weight 207.04
Percent Purity 95%
Chemical Name or Material 4-(Isopropylcarbamoyl)benzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.