Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Isopropylpiperidine 98.0+%, TCI America™
SDP

Catalog No. I07331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07331G 1 g
I07335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07331G Supplier TCI America Supplier No. I07331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19678-58-1
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD09055342
InChI Key YBPWIUSXQXYTSR-UHFFFAOYSA-N
PubChem CID 140569
IUPAC Name 4-(propan-2-yl)piperidine
SMILES CC(C)C1CCNCC1

Specifications

Color Yellow
Boiling Point 171°C
Quantity 1 g
UN Number 1993
Formula Weight 127.23
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Isopropylpiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.