Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Mercaptobenzoic acid, 90%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1074-36-8
C7H6O2S
154.19
MFCD00016617
LMJXSOYPAOSIPZ-UHFFFAOYSA-N
4-mercaptobenzoic acid,4-mercaptobenzoate,benzoic acid, 4-mercapto,4-mercapto-benzoic acid,4-mercapto benzoic acid,4-thiobenzoic acid,p-mercaptobenzoic acid,benzoic acid, p-mercapto,para-mercaptobenzoate,paracarboxythiophenol
95738
4-sulfanylbenzoic acid
C1=CC(=CC=C1C(=O)O)S
1074-36-8
C7H6O2S
154.19
MFCD00016617
LMJXSOYPAOSIPZ-UHFFFAOYSA-N
4-mercaptobenzoic acid,4-mercaptobenzoate,benzoic acid, 4-mercapto,4-mercapto-benzoic acid,4-mercapto benzoic acid,4-thiobenzoic acid,p-mercaptobenzoic acid,benzoic acid, p-mercapto,para-mercaptobenzoate,paracarboxythiophenol
95738
4-sulfanylbenzoic acid
C1=CC(=CC=C1C(=O)O)S
Packaging Glass bottle
Melting Point 223.0°C to 225.0°C
Quantity 25g
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula HSC6H4CO2H
Formula Weight 154.19
Percent Purity 90%
Chemical Name or Material 4-Mercaptobenzoic acid

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning