Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-3-nitrobiphenyl 99.0+%, TCI America™
SDP

Catalog No. M116625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M116625G 25 g
M11665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M116625G Supplier TCI America Supplier No. M116625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15854-73-6
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.24
MDL Number MFCD00191554
InChI Key HVYOIIIEAVPMCR-UHFFFAOYSA-N
Synonym 2-Nitro-4-phenylanisole
PubChem CID 615238
IUPAC Name 4-methoxy-3-nitro-1,1'-biphenyl
SMILES COC1=C(C=C(C=C1)C1=CC=CC=C1)[N+]([O-])=O

Specifications

Melting Point 89°C
Color Yellow
Quantity 25 g
Formula Weight 229.24
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxy-3-nitrobiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.