Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methoxy-N-methylformanilide 97.0+%, TCI America™
SDP

Catalog No. M17421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17421G 1 g
M17425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17421G Supplier TCI America Supplier No. M17421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5279-51-6
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.192
InChI Key AOXIKOMIXYQQLL-UHFFFAOYSA-N
Synonym N-(4-Methoxyphenyl)-N-methylformamide
PubChem CID 11309718
IUPAC Name N-(4-methoxyphenyl)-N-methylformamide
SMILES CN(C=O)C1=CC=C(C=C1)OC

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 165.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4′-Methoxy-N-methylformanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.