Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-TEMPO, free radical, 98+%, Thermo Scientific™

Catalog No. AAL1591503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1591503 1 g
AAL1591506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1591503 Supplier Thermo Scientific Chemicals Supplier No. L1591503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95407-69-5
Molecular Formula C10H20NO2
MDL Number MFCD00270334
Synonym 4-Methoxy-2,2,6,6-tetramethylpiperidinyloxy, free radical

Specifications

Melting Point 39°C to 41°C
Quantity 1 g
Beilstein 4740399
Formula Weight 186.28
Percent Purity ≥98%
Chemical Name or Material 4-Methoxy-TEMPO, free radical
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.