Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzenediazonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. M263625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M263625G 25 g
M26365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M263625G Supplier TCI America Supplier No. M263625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 459-64-3
Molecular Formula C7H7BF4N2O
Molecular Weight (g/mol) 221.95
MDL Number MFCD00011897
InChI Key CACFTKIREZJSIG-UHFFFAOYSA-J
Synonym 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide
PubChem CID 517233
IUPAC Name boron(3+) 4-methoxybenzene-1-diazonium tetrafluoride
SMILES [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point 139°C
Quantity 25 g
UN Number 1759
Formula Weight 221.95
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzenediazonium Tetrafluoroborate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.