Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzohydrazide 98.0+%, TCI America™
SDP

Catalog No. M177025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M177025G 25 g
M17705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M177025G Supplier TCI America Supplier No. M177025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3290-99-1
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017073
InChI Key REKQLYUAUXYJSZ-UHFFFAOYSA-N
Synonym 4-methoxybenzhydrazide, p-anisohydrazide, p-anisic hydrazide, p-anisoylhydrazine, 4-methoxybenzoic acid hydrazide, 4-methoxybenzoylhydrazine, anisoylhydrazine, anisic hydrazide, 4-methoxybenzene-1-carbohydrazide, p-anisic acid, hydrazide
PubChem CID 76792
IUPAC Name 4-methoxybenzohydrazide
SMILES COC1=CC=C(C=C1)C(=O)NN

Specifications

Melting Point 138°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxybenzohydrazide
RTECSNumber BZ4800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.