Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxybenzonitrile, 99%

Catalog No. AAA1594514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1594514 25 g
AAA1594506 5 g
AAA1594522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1594514 Supplier Thermo Scientific Chemicals Supplier No. A1594514
Add to Cart

CAS: 874-90-8 | C8H7NO | 133.15 g/mol

4-Methoxybenzonitrile is used as raw material of parfum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzonitrile is used as raw material of parfum.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-90-8
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001818
InChI Key XDJAAZYHCCRJOK-UHFFFAOYSA-N
Synonym anisonitrile, p-anisonitrile, 4-cyanoanisole, benzonitrile, 4-methoxy, p-methoxybenzonitrile, 4-anisonitrile, p-anisylnitrile, p-cyanoanisole, p-methoxyphenyl cyanide, 4-methoxy benzonitrile
PubChem CID 70129
IUPAC Name 4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1)C#N

Specifications

Melting Point 57°C to 61°C
Boiling Point 256°C to 257°C
Odor Almond-like
Quantity 25 g
UN Number UN3439
Beilstein 508117
Solubility Information Insoluble in water.
Formula Weight 133.15
Percent Purity 99%
Chemical Name or Material 4-Methoxybenzonitrile
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 212-871-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.